missing translation for 'onlineSavingsMsg'
Learn More

2-Propanethiol, 98%

Product Code. 10316780 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 mL
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
2.5 L
500mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10316780 500 mL Glass bottle 500mL
10071420 50 mL Glass bottle 50mL
10475165 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10316780 Supplier Thermo Scientific Acros Supplier No. 131445000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 75-33-2 | C3H8S | 76.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-33-2
Molecular Formula C3H8S
Molecular Weight (g/mol) 76.15
MDL Number MFCD00004863
InChI Key KJRCEJOSASVSRA-UHFFFAOYSA-N
Synonym 2-propanethiol, isopropyl mercaptan, isopropanethiol, isopropylthiol, 2-mercaptopropane, 2-propylmercaptan, 1-methylethanethiol, isopropylmercaptan, 2-propylthiol, 2-propane thiol
PubChem CID 6364
ChEBI CHEBI:8474
IUPAC Name propane-2-thiol
SMILES CC(C)S

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -131°C
Density 0.8200g/mL
Boiling Point 57°C to 60°C
Flash Point -34°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2CHSH
Refractive Index 1.4245 to 1.4265
Quantity 500 mL
Beilstein 01, 367
Specific Gravity 0.82
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohols,ethyl ether,hydrocarbons
Formula Weight 76.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Propanethiol, 98%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Propanethiol
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Flammable liquid Category 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.