Learn More
2-Propanethiol, 98%, Acros Organics
Brand: Acros Organics 131440500
Code : 29
Additional Details : CAS Number : 75-33-2 Weight : 0.09000kg Transport : UN number : 2402 Chem class : 3 Pack group : II
Packaging | Glass bottle |
---|---|
Quantity | 50mL |
Chemical Identifiers
75-33-2 | |
76.15 | |
KJRCEJOSASVSRA-UHFFFAOYSA-N | |
6364 | |
propane-2-thiol |
C3H8S | |
MFCD00004863 | |
2-propanethiol, isopropyl mercaptan, isopropanethiol, isopropylthiol, 2-mercaptopropane, 2-propylmercaptan, 1-methylethanethiol, isopropylmercaptan, 2-propylthiol, 2-propane thiol | |
CHEBI:8474 | |
CC(C)S |
Specifications
2-Propanethiol | |
-34°C | |
Glass bottle | |
0.82 | |
Undesignated | |
50mL | |
75-33-2 | |
100.0 | |
C3H8S | |
MFCD00004863 | |
2-propanethiol, isopropyl mercaptan, isopropanethiol, isopropylthiol, 2-mercaptopropane, 2-propylmercaptan, 1-methylethanethiol, isopropylmercaptan, 2-propylthiol, 2-propane thiol | |
KJRCEJOSASVSRA-UHFFFAOYSA-N | |
propane-2-thiol | |
6364 | |
76.15 | |
≥97.5% |
0.8200g/mL | |
Authentic | |
1.4245 to 1.4265 | |
57°C to 60°C | |
-131°C | |
98% | |
97.5 | |
98% | |
(CH3)2CHSH | |
01, 367 | |
Solubility in water: slightly soluble. Other solubilities: soluble in alcohols,ethyl ether,hydrocarbons | |
CC(C)S | |
76.15 | |
CHEBI:8474 | |
Liquid |
Safety and Handling
- 2-Propanethiol
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Flammable liquid Category 2
- Respiratory sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.