missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Propane-d7-thiol, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 2-Propanethiol-D7 |
| CAS | 1219803-56-1 |
| Molecular Formula | C3 D7 H S |
| Quantity | 0.25 g |
| Synonym | 2-Propane-d7-thiol, 2-Propanethiol-d7, 2-Propanethiol-D7, 1-(Methyl-d3)ethane-1,2,2,2-d4-thiol, 2-Mercaptopropane-d7, 2-Propylmercaptan-d7, Isopropylthiol-d7 |
| InChI Formula | InChI=1 S/C3H8S/c1-3(2)4/h3-4 H,1-2H3/i1D3,2D3,3 D |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 1,1,1,2,3,3,3-heptadeuteriopropane-2-thiol |
| Molecular Weight (g/mol) | 83.2038 |
| Formula Weight | 83.0786 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction