missing translation for 'onlineSavingsMsg'
Learn More

2-PMPA, MedChemExpress

Product Code. 16459716
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16459716

Brand: Medchem Express HY100788

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 2-PMPA is a potent and selective inhibitor of glutamate carboxypeptidase II (GCPII) with an IC50 of 300 pM.

2-PMPA is a potent and selective inhibitor of glutamate carboxypeptidase II (GCPII) with an IC50 of 300 pM.

Specifications

Chemical Name or Material 2-PMPA
Physical Form Viscous Oil
CAS 173039-10-6
Color Light Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C6H11O7P
For Use With (Application) Neuroscience-Neuromodulation
Synonym 2-(Phosphonomethyl)pentanedioic acid
Solubility Information H2O : ≥ 28 mg/mL (123.83 mM)
SMILES O=C(O)C(CP(O)(O)=O)CCC(O)=O
Molecular Weight (g/mol) 226.12
Formula Weight 226.12
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.