missing translation for 'onlineSavingsMsg'
Learn More

2-(Piperidylmethyl)-1-cyclohexanone hydrochloride, 98%, Thermo Scientific™

Catalog Number p-185320
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
6966-09-2
MFCD00068799
MFMOSYZXKAREGX-UHFFFAOYNA-N
pimeclone hydrochloride, 2-piperidylmethyl-1-cyclohexanone hydrochloride, karion respiratory stimulant, 1-piperidinomethyl-2-cyclohexanone hydrochloride, 2-1-piperidinylmethyl cyclohexanone hydrochloride, 2-piperidin-1-ylmethyl cyclohexanone hydrochloride, cyclohexanone, 2-piperidinomethyl-, hydrochloride, 2-piperidin-1-ylmethyl cyclohexanone, pimeclone hcl, 2-n-piperidinomethyl cyclohexanone hydrochloride
197860
This item is not returnable. View return policy
6966-09-2
MFCD00068799
MFMOSYZXKAREGX-UHFFFAOYNA-N
pimeclone hydrochloride, 2-piperidylmethyl-1-cyclohexanone hydrochloride, karion respiratory stimulant, 1-piperidinomethyl-2-cyclohexanone hydrochloride, 2-1-piperidinylmethyl cyclohexanone hydrochloride, 2-piperidin-1-ylmethyl cyclohexanone hydrochloride, cyclohexanone, 2-piperidinomethyl-, hydrochloride, 2-piperidin-1-ylmethyl cyclohexanone, pimeclone hcl, 2-n-piperidinomethyl cyclohexanone hydrochloride
197860

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 166°C to 170°C
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C12H21NO·HCl
Quantity 1 g
Merck Index 13, 7510
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in methanol,ethanol
Molecular Weight (g/mol) 231.77
Formula Weight 231.77
Percent Purity 98%
Chemical Name or Material 2-(Piperidylmethyl)-1-cyclohexanone hydrochloride, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'