missing translation for 'onlineSavingsMsg'
Learn More

2-Piperidinoisonicotinic acid, 97%, Thermo Scientific™

Product Code. 10081094
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10081094 1 g Amber Glass Bottle 1g
10143504 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10081094 Supplier Thermo Scientific Maybridge Supplier No. CC50301DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 855153-75-2
Molecular Formula C11H14N2O2
Molecular Weight (g/mol) 206.245
MDL Number MFCD04115214
InChI Key PYHYIVXTQSCHDX-UHFFFAOYSA-N
Synonym 2-piperidinoisonicotinic acid, 2-piperidin-1-yl isonicotinic acid, 2-piperidin-1-yl pyridine-4-carboxylic acid, 2-piperidin-1-ylisonicotinic acid, 2-piperidin-1-ylpyridin-4-ylcarboxylic acid, 3,4,5,6-tetrahydro-2h-1,2' bipyridinyl-4'-carboxylic acid, 4-pyridinecarboxylic acid, 2-1-piperidinyl, 2-piperidylpyridine-4-carboxylic acid, 2-piperidin-1-yl-isonicotinic acid, 1-4-carboxypyridin-2-yl piperidine
PubChem CID 6484237
IUPAC Name 2-piperidin-1-ylpyridine-4-carboxylic acid
SMILES C1CCN(CC1)C2=NC=CC(=C2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 206.24
Percent Purity 97%
Chemical Name or Material 2-Piperidinoisonicotinic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.