Learn More
2-piperidinobenzamide, 97%, Maybridge™
Brand: Maybridge BTB03853DA
Code : 31
Additional Details : CAS Number : 3430-40-8 Weight : 0.05100kg
Quantity | 1g |
---|
Chemical Identifiers
3430-40-8 | |
204.273 | |
VTXYPPVXMJMLCY-UHFFFAOYSA-N | |
335067 | |
C1CCN(CC1)C2=CC=CC=C2C(=O)N |
C12H16N2O | |
MFCD00052257 | |
2-piperidinobenzamide, 2-piperidin-1-yl benzamide, benzamide,2-1-piperidinyl, 2-piperidylbenzamide, piperidinyl benzamide, maybridge1_000974, 2-1-piperidinyl benzamide, benzamide, 2-1-piperidinyl | |
2-piperidin-1-ylbenzamide |
Specifications
2-Piperidinobenzamide | |
1g | |
96.5 | |
97% | |
MFCD00052257 | |
VTXYPPVXMJMLCY-UHFFFAOYSA-N | |
2-piperidin-1-ylbenzamide | |
335067 | |
97% |
Amber glass bottle | |
3430-40-8 | |
100.0 | |
C12H16N2O | |
2-piperidinobenzamide, 2-piperidin-1-yl benzamide, benzamide,2-1-piperidinyl, 2-piperidylbenzamide, piperidinyl benzamide, maybridge1_000974, 2-1-piperidinyl benzamide, benzamide, 2-1-piperidinyl | |
C1CCN(CC1)C2=CC=CC=C2C(=O)N | |
204.273 | |
204.27 |