missing translation for 'onlineSavingsMsg'
Learn More

2-Piperidineethanol, 95%

Product Code. 10163300 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10163300 100 g Glass bottle 100g
10474725 5 g Glass bottle 5g
10031710 500 g Glass bottle 500g
11946301 2.5 kg Glass bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10163300 Supplier Thermo Scientific Acros Supplier No. 157611000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1484-84-0 | C7H15NO | 129.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1484-84-0
Molecular Formula C7H15NO
Molecular Weight (g/mol) 129.2
InChI Key PTHDBHDZSMGHKF-UHFFFAOYSA-N
Synonym 2-piperidineethanol, 2-2-hydroxyethyl piperidine, 2-piperidin-2-yl ethanol, 2-2-piperidyl ethan-1-ol, 2-piperidine ethanol, 2-hydroxyethyl piperidine, 2-piperidine-ethanol, piperidine-2-ethanol, 2-piperidin-2-yl ethan-1-ol
PubChem CID 15144
IUPAC Name 2-piperidin-2-ylethanol
SMILES C1CCNC(C1)CCO

Specifications

Melting Point 38.0°C to 43.0°C
Color White to Yellow
Density 1.0100g/mL
Boiling Point 234.0°C
Flash Point 102°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 100 g
Beilstein 21,2
Specific Gravity 1.01
Solubility Information Solubility in water: very soluble. Other solubilities: soluble in alcohol
Formula Weight 129.2
Percent Purity 95%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material 2-Piperidineethanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Piperidineethanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.