missing translation for 'onlineSavingsMsg'
Learn More

2-Piperazinone, TRC

Product Code. 30414192
Change view
Click to view available options
Quantity:
2 g
Unit Size:
2g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30414192 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30414192 Supplier Toronto Research Chemicals Supplier No. TRCP4801402G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Piperazinone
CAS 5625-67-2
Molecular Formula C4H8N2O
Quantity 2 g
Synonym Tetrahydro-2(1H)-pyrazinone, 2-Ketopiperazine, 2-Oxopiperazine, 3-Oxopiperazine, Oxopiperazine, Piperazin-2-one, Piperazine-2-one, NSC 27441
InChI Formula InChI=1S/C4H8N2O/c7-4-3-5-1-2-6-4/h5H,1-3H2,(H,6,7)
SMILES O=C1CNCCN1
IUPAC Name piperazin-2-one
Molecular Weight (g/mol) 100.12
Formula Weight 100.0637
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.