missing translation for 'onlineSavingsMsg'
Learn More

2-Picolinic acid, 99%

Product Code. 11359559
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11359559 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11359559 Supplier Thermo Scientific Alfa Aesar Supplier No. A14749.14
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 98-98-6 | C6H5NO2 | 123.11 g/mol

2-Picolinic acid is used as a chelate for alkaline earth metals. Used to prepare picolinato ligated transition metal complexes. In synthetic organic chemistry, has been used as a substrate in the Mitsunobu reaction and in the Hammick reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Picolinic acid is used as a chelate for alkaline earth metals. Used to prepare picolinato ligated transition metal complexes. In synthetic organic chemistry, has been used as a substrate in the Mitsunobu reaction and in the Hammick reaction.

Solubility
Soluble in water. Also soluble in glacial acetic acid, ethanol and methanol. Insoluble in chloroform.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, reducing agents and bases.
TRUSTED_SUSTAINABILITY

Specifications

CAS 98-98-6
Melting Point 135°C to 139°C
Flash Point 139°C
Odor Odorless
Molecular Formula C6H5NO2
MDL Number MFCD00006293
Quantity 25 g
Beilstein 109595
Merck Index 14,7403
Synonym picolinic acid, 2-pyridinecarboxylic acid, 2-picolinic acid, 2-carboxypyridine, alpha-picolinic acid
Solubility Information Soluble aq. soln.. Also soluble in glacial acetic acid,ethanol and methanol. Insoluble in chloroform.
InChI Key SIOXPEMLGUPBBT-UHFFFAOYSA-N
SMILES OC(=O)C1=CC=CC=N1
IUPAC Name pyridine-2-carboxylic acid
Molecular Weight (g/mol) 123.11
PubChem CID 1018
ChEBI CHEBI:28747
Formula Weight 123.11
Percent Purity 99%
Chemical Name or Material 2-Picolinic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.