missing translation for 'onlineSavingsMsg'
Learn More

2-Picolinic acid, 99%

Product Code. 11477796
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11477796 500 g 500g
11467796 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11477796 Supplier Thermo Scientific Alfa Aesar Supplier No. A14749.36
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 98-98-6 | C6H5NO2 | 123.111 g/mol

2-Picolinic acid is used as a chelate for alkaline earth metals. Used to prepare picolinato ligated transition metal complexes. In synthetic organic chemistry, has been used as a substrate in the Mitsunobu reaction and in the Hammick reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Picolinic acid is used as a chelate for alkaline earth metals. Used to prepare picolinato ligated transition metal complexes. In synthetic organic chemistry, has been used as a substrate in the Mitsunobu reaction and in the Hammick reaction.

Solubility
Soluble in water. Also soluble in glacial acetic acid, ethanol and methanol. Insoluble in chloroform.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents, reducing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-98-6
Molecular Formula C6H5NO2
Molecular Weight (g/mol) 123.111
MDL Number MFCD00006293
InChI Key SIOXPEMLGUPBBT-UHFFFAOYSA-N
Synonym picolinic acid, 2-pyridinecarboxylic acid, 2-picolinic acid, 2-carboxypyridine, alpha-picolinic acid, o-pyridinecarboxylic acid, pyridinecarboxylic acid, alpha-pyridinecarboxylic acid, picolinate, acide picolique
PubChem CID 1018
ChEBI CHEBI:28747
IUPAC Name pyridine-2-carboxylic acid
SMILES C1=CC=NC(=C1)C(=O)O

Specifications

Melting Point 135°C to 139°C
Flash Point 139°C (232°F)
Odor Odorless
Quantity 500 g
Beilstein 109595
Merck Index 14,7403
Solubility Information Soluble in water. Also soluble in glacial acetic acid,ethanol and methanol. Insoluble in chloroform.
Formula Weight 123.11
Percent Purity 99%
Chemical Name or Material 2-Picolinic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.