missing translation for 'onlineSavingsMsg'
Learn More

2-Picoline, 98+%

Product Code. 11429822
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2500mL
500mL
This item is not returnable. View return policy

Product Code. 11429822

Brand: Thermo Scientific Alfa Aesar A10847.0F

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 109-06-8 | C6H7N | 93.129 g/mol

2-Picoline is used as an intermediate in agrochemicals and pharmaceuticals. It serves as a solvent as well as to prepare dyes and resins. It finds application as a constituent in cigarette smoke, bone oil, coal tar and coke oven emissions. Further, it acts as a precursor of 2-vinylpyridine, picolinic acid and nitrapyrin. It is also employed to study the electron and proton transfer reactions of lumiflavin. In addition, it is used in the synthetic pathway for the preparation of dearomatized, allylated and carbon-hydrogen bond activated pyridine derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Picoline is used as an intermediate in agrochemicals and pharmaceuticals. It serves as a solvent as well as to prepare dyes and resins. It finds application as a constituent in cigarette smoke, bone oil, coal tar and coke oven emissions. Further, it acts as a precursor of 2-vinylpyridine, picolinic acid and nitrapyrin. It is also employed to study the electron and proton transfer reactions of lumiflavin. In addition, it is used in the synthetic pathway for the preparation of dearomatized, allylated and carbon-hydrogen bond activated pyridine derivatives.

Solubility
Miscible with water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents, sulfuric acid, acids, metals, iron(II) sulfate and hydrogen peroxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-06-8
Molecular Formula C6H7N
Molecular Weight (g/mol) 93.129
MDL Number MFCD00006332
InChI Key BSKHPKMHTQYZBB-UHFFFAOYSA-N
Synonym 2-picoline, o-picoline, pyridine, 2-methyl, picoline, alpha-picoline, a-picoline, pyridine, methyl, methylpyridine, o-methylpyridine, picoline, alpha
PubChem CID 7975
ChEBI CHEBI:50415
IUPAC Name 2-methylpyridine
SMILES CC1=CC=CC=N1

Specifications

Melting Point -70°C
Density 0.944
Boiling Point 128°C to 129°C
Flash Point 26°C (78°F)
Refractive Index 1.5
Quantity 2500 mL
UN Number UN2313
Beilstein 104581
Merck Index 14,7400
Solubility Information Miscible with water.
Formula Weight 93.13
Percent Purity ≥98%
Chemical Name or Material 2-Picoline
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.