missing translation for 'onlineSavingsMsg'
Learn More

2-(Phthalimido)ethanesulfonyl chloride, 97%

Product Code. p-7033288
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11315369 1 g 1g
11325369 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11315369 Supplier Thermo Scientific Alfa Aesar Supplier No. H50382.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4403-36-5 | C10H8ClNO4S | 273.687 g/mol

2-(Phthalimido)ethanesulfonyl chloride is an important raw material and intermediate used in pharmaceuticals, agro based and dye stuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Phthalimido)ethanesulfonyl chloride is an important raw material and intermediate used in pharmaceuticals, agro based and dye stuff fields.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4403-36-5
Molecular Formula C10H8ClNO4S
Molecular Weight (g/mol) 273.687
MDL Number MFCD01861217
InChI Key HCPVYBCAYPMANM-UHFFFAOYSA-N
Synonym 2-phthalimidoethanesulfonyl chloride, 2-1,3-dioxoisoindolin-2-yl ethanesulfonylchloride, 2-1,3-dioxoisoindolin-2-yl ethanesulfonyl chloride, 2-1,3-dioxo-1,3-dihydro-2h-isoindol-2-yl ethanesulfonyl chloride, 2-phthalimidoethanesulfonylchloride, 2-phthalimido ethanesulfonyl chloride, 2-1,3-dioxoisoindol-2-yl ethanesulfonyl chloride, 2-1,3-dioxo-2,3-dihydro-1h-isoindol-2-yl ethane-1-sulfonyl chloride, 2-1,3-dioxo-1,3-dihydro-isoindol-2-yl-ethanesulfonyl chloride, 2-2-chlorosulfonyl ethyl benzo c azoline-1,3-dione
PubChem CID 308739
IUPAC Name 2-(1,3-dioxoisoindol-2-yl)ethanesulfonyl chloride
SMILES C1=CC=C2C(=C1)C(=O)N(C2=O)CCS(=O)(=O)Cl

Specifications

Melting Point 156°C to 160°C
Quantity 1 g
UN Number UN3261
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 273.7
Percent Purity 97%
Chemical Name or Material 2-(Phthalimido)ethanesulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.