missing translation for 'onlineSavingsMsg'
Learn More

2-Phospho-L-ascorbic Acid Trisodium Salt, TRC

Product Code. 30410268
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
Product Code. Quantity unitSize
30410268 10 g 10g
30414180 25 g -
2 options
This item is not returnable. View return policy

Product Code. 30410268

Brand: Toronto Research Chemicals TRCP35400310G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66170-10-3
Formula Weight 321.9443
InChI Formula InChI=1S/C6H9O9P.3Na/c7-1-2(8)4-3(9)5(6(10)14-4)15-16(11,12)13;;;/h2,4,7-9H,1H2,(H2,11,12,13);;;/q;3*+1/p-3/t2-,4+;;;/m0.../s1
IUPAC Name trisodium;[(2R)-2-[(1S)-1,2-dihydroxyethyl]-3-oxido-5-oxo-2H-furan-4-yl] phosphate
Molecular Formula C6H6Na3O9P
Molecular Weight (g/mol) 322.05
SMILES [Na+].[Na+].[Na+].OC[C@H](O)[C@H]1OC(=O)C(=C1[O-])OP(=O)([O-])[O-]
Synonym Ascorbic Acid Phosphate Sodium Salt, L-Ascorbic Acid 2-(Dihydrogen Phosphate) Trisodium Salt, Sodium Ascorbyl Phosphate, Stay-C 50, Trisodium Ascorbate-2-phosphate, VCP-NA
Chemical Name or Material 2-Phospho-L-ascorbic Acid Trisodium Salt

Specifications

Quantity 10 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.