missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylthiophene, 95%

Product Code. 15477318
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15477318

Brand: Thermo Scientific Alfa Aesar H60819.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 825-55-8 | C10H8S | 160.234 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 825-55-8
Molecular Formula C10H8S
Molecular Weight (g/mol) 160.234
MDL Number MFCD00130080
InChI Key PJRGDKFLFAYRBV-UHFFFAOYSA-N
Synonym thiophene, 2-phenyl, 2-phenyl-thiophene, phenylthiophene, thiophene, phenyl, 2-thienylbenzene, 2-phenyithiophene, 2-phenylthiofuran, 2-phenylthiophene, acmc-209ppe
PubChem CID 69999
IUPAC Name 2-phenylthiophene
SMILES C1=CC=C(C=C1)C2=CC=CS2

Specifications

Melting Point 34°C to 36°C
Boiling Point 256°C
Flash Point 109°C (228°F)
Quantity 1 g
Formula Weight 160.24
Percent Purity 95%
Chemical Name or Material 2-Phenylthiophene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.