missing translation for 'onlineSavingsMsg'
Learn More

2-(Phenylthio)nicotinic acid, 97%, Thermo Scientific™

Product Code. 10334512
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10334512 10 g Amber Glass Bottle 10g
10552092 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10334512 Supplier Thermo Scientific Maybridge Supplier No. BTB08722EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 35620-72-5
Molecular Formula C12H9NO2S
Molecular Weight (g/mol) 231.269
MDL Number MFCD00052110
InChI Key MGIDOIDQQMOYTH-UHFFFAOYSA-N
Synonym 2-phenylthio nicotinic acid, 2-phenylsulfanyl nicotinic acid, 2-phenylsulfanyl pyridine-3-carboxylic acid, 3-pyridinecarboxylicacid, 2-phenylthio, 3-pyridinecarboxylic acid, 2-phenylthio, 2-phenylthiopyridine-3-carboxylic acid, maybridge1_001786, 2-phenylthio-3-pyridinecarboxylic acid, 2-phenylthio pyridine-3-carboxylic acid
PubChem CID 725029
IUPAC Name 2-phenylsulfanylpyridine-3-carboxylic acid
SMILES C1=CC=C(C=C1)SC2=C(C=CC=N2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 231.27
Percent Purity 97%
Chemical Name or Material 2-(Phenylthio)nicotinic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.