missing translation for 'onlineSavingsMsg'
Learn More

2-(Phenylthio)ethanol, 99%, Thermo Scientific Chemicals

Product Code. 11479842
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11479842 10 g 10g
11489842 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11479842 Supplier Thermo Scientific Alfa Aesar Supplier No. A10861.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 699-12-7
Molecular Formula C8H10OS
Molecular Weight (g/mol) 154.23
MDL Number MFCD00002905
InChI Key KWWZHCSQVRVQGF-UHFFFAOYSA-N
Synonym 2-phenylthio ethanol, phenylthioethanol, ethanol, 2-phenylthio, 2-hydroxyethyl phenyl sulfide, 2-phenylmercaptoethanol, 2-phenylsulfanyl ethanol, 2-phenylthioethanol, unii-9wa374o44p, pubchem10784
PubChem CID 69685
IUPAC Name 2-phenylsulfanylethanol
SMILES OCCSC1=CC=CC=C1

Specifications

Density 1.147
Boiling Point 115°C to 116°C (2 mmHg)
Flash Point >110°C (230°F)
Odor Unpleasant
Refractive Index 1.592
Quantity 10 g
Beilstein 1860057
Formula Weight 154.23
Percent Purity 99%
Chemical Name or Material 2- (Phenylthio)ethanol
compliance-icons
Product Identifier
  • 2-(Phenylthio)ethanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 2-(Phenylthio)ethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.