missing translation for 'onlineSavingsMsg'
Learn More

2-(Phenylthio)ethanimidamide hydrochloride, 97%, Thermo Scientific™

Product Code. 10325492
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10325492 10 g 10g
10253572 1 g 1g
10757305 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10325492 Supplier Thermo Scientific Maybridge Supplier No. SPB05877EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 84544-86-5
Molecular Formula C8H11N2S
Molecular Weight (g/mol) 167.25
MDL Number MFCD00100031
InChI Key QTFCJAKFFGFURL-UHFFFAOYSA-O
Synonym 2-phenylthio ethanimidamide hydrochloride, 2-phenylthio acetamidine hydrochloride, 2-phenylsulfanyl ethanimidamide hydrochloride, 2-phenylsulfanylethanimidamide hydrochloride, 2-phenylthioethanamidine, chloride, 2-phenylthio acetimidamide hydrochloride
PubChem CID 2746028
IUPAC Name 2-phenylsulfanylethanimidamide;hydrochloride
SMILES NC(=[NH2+])CSC1=CC=CC=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Linear Formula C6H5SCH2C(=NH)NH2·HCl
Quantity 10 g
Formula Weight 202.71
Percent Purity 97%
Chemical Name or Material 2-(Phenylthio)ethanimidamide hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.