missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylthieno[3,2-c]pyridin-4(5H)-one, TRC

Product Code. 30426181
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
Product Code. Quantity unitSize
30426181 5 mg -
30426062 50 mg 50mg
30426061 25 mg 25mg
3 options
This item is not returnable. View return policy

Product Code. 30426181

Brand: Toronto Research Chemicals TRCP1203105MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 690636-03-4
Formula Weight 227.0405
InChI Formula InChI=1S/C13H9NOS/c15-13-10-8-12(9-4-2-1-3-5-9)16-11(10)6-7-14-13/h1-8H,(H,14,15)
IUPAC Name 2-phenyl-5H-thieno[3,2-c]pyridin-4-one
Molecular Formula C13H9NOS
Molecular Weight (g/mol) 227.29
SMILES O=C1NC=Cc2sc(cc12)c3ccccc3
Synonym 2-Phenyl-4-oxo-4,5-dihydrothieno[3,2-c]pyridine
Chemical Name or Material 2-Phenylthieno[3,2-c]pyridin-4(5H)-one

Specifications

Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.