missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylthiazolidine-4-carboxylic acid, 97%, Thermo Scientific™

Product Code. 15469218
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42607-21-6
Molecular Formula C10H11NO2S
Molecular Weight (g/mol) 209.263
MDL Number MFCD00170683
InChI Key AZDYQBFYMBALBY-UHFFFAOYSA-N
Synonym 2-phenylthiazolidine-4-carboxylic acid, 2-phenyl-4-thiazolidinecarboxylic acid, 4-carboxy-2-phenylthiazolidine, 2-phenyl-1,3-thiazolane-4-carboxylic acid, 4-thiazolidinecarboxylic acid, 2-phenyl, 4-thiazolidinecarboxylic acid 2-phenyl, 2-phenyl-thiazolidine-4-carboxylic acid, 2-phenyl-4-thiazolidine-4-carboxylic acid, phenylthiazolanecarboxylicacid, #
PubChem CID 39237
IUPAC Name 2-phenyl-1,3-thiazolidine-4-carboxylic acid
SMILES C1C(NC(S1)C2=CC=CC=C2)C(=O)O

Specifications

Color White
Quantity 1 g
Formula Weight 209.26
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Phenylthiazolidine-4-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'