missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylpyrimidine-5-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10032514
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10032514 1 g 1g
10728465 250 mg 250mg
10163214 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10032514 Supplier Thermo Scientific Maybridge Supplier No. CC66301DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 122773-97-1
Molecular Formula C11H8N2O2
Molecular Weight (g/mol) 200.197
MDL Number MFCD00559612
InChI Key BOAIYSRFGWBZCF-UHFFFAOYSA-N
Synonym 2-phenyl-5-pyrimidinecarboxylic acid, 5-carboxy-2-phenylpyrimidine, 5-pyrimidinecarboxylic acid, 2-phenyl, 2-phenyl-pyrimidine-5-carboxylic acid, 5-pyrimidinecarboxylicacid, 2-phenyl, acmc-1c8my, 5-carboxy-2-phenyl-1,3-diazine, 2-phenyl pyrimidine-5-carboxylic acid
PubChem CID 747389
IUPAC Name 2-phenylpyrimidine-5-carboxylic acid
SMILES C1=CC=C(C=C1)C2=NC=C(C=N2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 200.2
Percent Purity 97%
Chemical Name or Material 2-Phenylpyrimidine-5-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.