missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylpyrimidine-5-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10376654
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10376654 1 g Amber Glass Bottle 1g
10793834 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10376654 Supplier Thermo Scientific Maybridge Supplier No. CC66304DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 130161-46-5
Molecular Formula C11H8N2O
Molecular Weight (g/mol) 184.198
MDL Number MFCD03085924
InChI Key AUTGLFSBJLERMV-UHFFFAOYSA-N
Synonym 2-phenyl-5-pyrimidinecarboxaldehye, 2-phenylpyrimidine-5-carboxaldehyde, 5-pyrimidinecarboxaldehyde, 2-phenyl, 2-phenyl-pyrimidine-5-carbaldehyde, 5-pyrimidinecarboxaldehyde,2-phenyl, 2-phenyl-5-pyrimidinecarbaldehyde, 2-phenyl-5-pyrimidinecarboxaldehyde, acmc-1c230, 1-2-phenylpyrimidin-5-yl methanone
PubChem CID 820135
IUPAC Name 2-phenylpyrimidine-5-carbaldehyde
SMILES C1=CC=C(C=C1)C2=NC=C(C=N2)C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 184.2
Percent Purity 97%
Chemical Name or Material 2-Phenylpyrimidine-5-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.