missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylpropylamine hydrochloride, ≥98%, Thermo Scientific™

Product Code. 15477647
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20388-87-8
Molecular Formula C9H14ClN
Molecular Weight (g/mol) 171.668
MDL Number MFCD01708026
InChI Key HBVYOCJBEXSCQE-UHFFFAOYSA-N
Synonym 2-phenylpropan-1-amine hydrochloride, beta-methylphenethylamine hydrochloride, 2-phenyl-1-propylamine hcl, phenethylamine, beta-methyl-, hydrochloride, +-, dl-beta-phenyl-n-propylamine hydrochloride, phenethylamine, beta-methyl-, hydrochloride, +--beta-methylphenethylamine hydrochloride, 2-phenylpropylamine hydrochloride, 2-phenylpropan-1-amine hcl, 2-phenylpropan-1-amine hydrochloride 1:1
PubChem CID 89332
IUPAC Name 2-phenylpropan-1-amine;hydrochloride
SMILES CC(CN)C1=CC=CC=C1.Cl

Specifications

Melting Point 143°C to 145°C
Color Cream to Yellow
Quantity 5 g
Sensitivity Hygroscopic
Formula Weight 171.67
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material 2-Phenylpropylamine hydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'