missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylpropylamine hydrochloride, ≥98%, Thermo Scientific™

Product Code. p-7052547
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 15477647

Brand: Thermo Scientific Alfa Aesar H36338.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20388-87-8
Molecular Formula C9H14ClN
Molecular Weight (g/mol) 171.668
MDL Number MFCD01708026
InChI Key HBVYOCJBEXSCQE-UHFFFAOYSA-N
Synonym 2-phenylpropan-1-amine hydrochloride, beta-methylphenethylamine hydrochloride, 2-phenyl-1-propylamine hcl, phenethylamine, beta-methyl-, hydrochloride, +-, dl-beta-phenyl-n-propylamine hydrochloride, phenethylamine, beta-methyl-, hydrochloride, +--beta-methylphenethylamine hydrochloride, 2-phenylpropylamine hydrochloride, 2-phenylpropan-1-amine hcl, 2-phenylpropan-1-amine hydrochloride 1:1
PubChem CID 89332
IUPAC Name 2-phenylpropan-1-amine;hydrochloride
SMILES CC(CN)C1=CC=CC=C1.Cl

Specifications

Melting Point 143°C to 145°C
Color Cream to Yellow
Quantity 5 g
Sensitivity Hygroscopic
Formula Weight 171.67
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material 2-Phenylpropylamine hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.