missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylpropionaldehyde, 97%

Product Code. 11470577
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11470577 500 g 500g
11460577 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11470577 Supplier Thermo Scientific Alfa Aesar Supplier No. B22436.36
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 93-53-8 | C9H10O | 134.178 g/mol

2-phenylpropionaldehyde to give acetophenone using 1 atm molecular oxygen as oxidant was catalyzed by Ce(IV), V(IV) and Bronsted acids. It is used to produce 3-hydroxy-2-methyl-2-phenyl-propionaldehyde. This reaction will need reagent water and K2CO3.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-phenylpropionaldehyde to give acetophenone using 1 atm molecular oxygen as oxidant was catalyzed by Ce(IV), V(IV) and Bronsted acids. It is used to produce 3-hydroxy-2-methyl-2-phenyl-propionaldehyde. This reaction will need reagent water and K2CO3.

Solubility
Soluble in most fixed oils, propylene glycol. Insoluble in glycerin.

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-53-8
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.178
MDL Number MFCD00006973
InChI Key IQVAERDLDAZARL-UHFFFAOYSA-N
Synonym 2-phenylpropionaldehyde, hydratropaldehyde, hydratropic aldehyde, hyacinthal, cumene aldehyde, hydratropa aldehyde, hydrotropic aldehyde, 2-phenyl-1-propanal, 2-phenyl propionaldehyde, aldehyd hydratropovy
PubChem CID 7146
IUPAC Name 2-phenylpropanal
SMILES CC(C=O)C1=CC=CC=C1

Specifikationer

Density 1.01
Boiling Point 92°C to 94°C (12 mmHg)
Flash Point 79°C (174°F)
Refractive Index 1.517
Quantity 500 g
Beilstein 4291321
Sensitivity Air Sensitive
Solubility Information Soluble in most fixed oils,propylene glycol. Insoluble in glycerin.
Formula Weight 134.18
Percent Purity 97%
Chemical Name or Material 2-Phenylpropionaldehyde
Visa mer Visa mindre

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.