missing translation for 'onlineSavingsMsg'
Learn More

(±)-2-Phenylpropanoic Acid, TRC

Product Code. 17922875
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17922875 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17922875 Supplier Toronto Research Chemicals Supplier No. TRCP3361705G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 492-37-5
Molecular Formula C9 H10 O2
Quantity 5 g
Synonym Benzeneacetic acid, α-methyl-, Hydratropic acid (6CI,7CI,8CI), α-Methylbenzeneacetic acid, (RS)-2-Phenylpropanoic acid, (RS)-2-Phenylpropionic acid, (±)-2-Phenylpropanoic acid, (±)-2-Phenylpropionic acid, (±)-Hydratropic acid, (±)-α-Methylphenylacetic acid, 2-Phenylpropanoic acid, 2-Phenylpropionic acid, DL-2-Phenylpropionic acid, NSC 245033, NSC 42872, dl-PPA, α-Methylphenylacetic acid, α-Phenylpropanoic acid, α-Phenylpropionic acid
InChI Formula InChI=1S/C9H10O2/c1-7(9(10)11)8-5-3-2-4-6-8/h2-7H,1H3,(H,10,11)
SMILES CC(C(=O)O)c1ccccc1
IUPAC Name 2-phenylpropanoic acid
Molecular Weight (g/mol) 150.17
Formula Weight 150.0681
Chemical Name or Material 2-Phenylpropanoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.