missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Phenylphenol, 99%
Click to view available options
Quantity:
250 g
500 g
2500 g
Unit Size:
2500g
250g
500g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Applications
2-Phenylphenol is used as a dye intermediate, plasticizer, rubber chemical and as a preservative in water-oil emulsions. It is used as an antimicrobial preservative in cosmetics and an antimicrobial additive in the manufacture of metalworking fluids, leather, adhesives and textiles. Further, it serves as a disinfectant in households, hospitals, nursing homes, farms, laundries, barber shops, veterinary equipment and food processing plants. It is also employed in the production of resins and rubber chemicals. In addition to this, it is used as a polymer material, stabilizer and flame retardant.
Solubility
Soluble in ethanol, acetone, benzene,sodium hydroxide, chloroform, acetonitrile, toluene, hexane, ligroin, ethyl ether, pyridine, ethylene glycol, isopropanol, glycol ethers and polyglycols.
Notes
Incompatible with strong oxidizing agents and strong bases.
2-Phenylphenol is used as a dye intermediate, plasticizer, rubber chemical and as a preservative in water-oil emulsions. It is used as an antimicrobial preservative in cosmetics and an antimicrobial additive in the manufacture of metalworking fluids, leather, adhesives and textiles. Further, it serves as a disinfectant in households, hospitals, nursing homes, farms, laundries, barber shops, veterinary equipment and food processing plants. It is also employed in the production of resins and rubber chemicals. In addition to this, it is used as a polymer material, stabilizer and flame retardant.
Solubility
Soluble in ethanol, acetone, benzene,sodium hydroxide, chloroform, acetonitrile, toluene, hexane, ligroin, ethyl ether, pyridine, ethylene glycol, isopropanol, glycol ethers and polyglycols.
Notes
Incompatible with strong oxidizing agents and strong bases.
Specifications
Specifications
| Melting Point | 54°C to 58°C |
| Density | 1.293 |
| Boiling Point | 152°C to 154°C (15 mmHg) |
| Flash Point | 124°C (255°F) |
| Odor | Mild |
| Quantity | 500 g |
| UN Number | UN3077 |
| Beilstein | 606907 |
| Merck Index | 14,7304 |
| Solubility Information | Soluble in ethanol,acetone,benzene,sodium hydroxide,chloroform,acetonitrile,toluene,hexane,ligroin,ethyl ether,pyridine,ethylene glycol,isopropanol,glycol ethers and polyglycols. |
| Show More |
Safety and Handling
Product Identifier
- 2-Phenylphenol
- Warning
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H400-Very toxic to aquatic life.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : 2-Phenylphenol
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction