missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylindole, 98%, Thermo Scientific Chemicals

Product Code. 10376820 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10376820 500 g Plastic bottle 500g
10081440 5 g Glass bottle 5g
10326920 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10376820 Supplier Thermo Scientific Acros Supplier No. 130685000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 948-65-2 | C14H11N | 193.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 948-65-2
Molecular Formula C14H11N
Molecular Weight (g/mol) 193.25
MDL Number MFCD00005608
InChI Key KLLLJCACIRKBDT-UHFFFAOYSA-N
Synonym 2-phenylindole, 1h-indole, 2-phenyl, indole, 2-phenyl, stabilizer i, alpha-phenylindole, 2-phenyl indole, phenylindole, unii-mqd44hv3p1, .alpha.-phenylindole, mqd44hv3p1
PubChem CID 13698
IUPAC Name 2-phenyl-1H-indole
SMILES C1=CC=C(C=C1)C2=CC3=CC=CC=C3N2

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 188.0°C to 191.0°C
Color Beige to White or Green
Boiling Point 250.0°C (10.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Plastic bottle
Quantity 500 g
Beilstein 20, 467
Solubility Information Solubility in water: .. Other solubilities: soluble in thf and acetone,slightly soluble in methanol
Formula Weight 193.25
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 2-Phenylindole, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.