missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylimidazole, 98%

Product Code. 10431592 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10431592 100 g Glass bottle 100g
10653172 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10431592 Supplier Thermo Scientific Acros Supplier No. 356431000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 670-96-2 | C9H8N2 | 144.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 670-96-2
Molecular Formula C9H8N2
Molecular Weight (g/mol) 144.18
MDL Number MFCD00005186
InChI Key ZCUJYXPAKHMBAZ-UHFFFAOYSA-N
Synonym 2-phenylimidazole, 1h-imidazole, 2-phenyl, imidazole, 2-phenyl, 2-phenyl imidazole, 2-phenyl-1h-imidazol, unii-7r2590hm4l, chembl14189, phenyl imidazole, 2-phenyl-imidazole, piy
PubChem CID 69591
IUPAC Name 2-phenyl-1H-imidazole
SMILES N1C=CN=C1C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 141.0°C to 148.0°C
Color Yellow to Beige
Boiling Point 340.0°C
Flash Point 200°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 100 g
Beilstein 23, 182
Solubility Information Solubility in water: insoluble
Formula Weight 144.18
Percent Purity 98%
Physical Form Crystalline Powder, Powder or Granular
Chemical Name or Material 2-Phenylimidazole, 95%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Phenylimidazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.