missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylethylboronic acid, 98%, Thermo Scientific™

Product Code. 15479247
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15479247

Brand: Thermo Scientific Alfa Aesar H52502.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Phenylethylboronic acid is used as a reactant in Suzuki-Miyaura cross-coupling reactions, reactions with α-diazocarbonyl compounds, C-H functionalization of quinones, cross-coupling with aromatic amines and arylation & alkylation of diphenylisoxazole. It has been also used as reactant in studies of the stability of boronic esters to hydrolysis.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34420-17-2
Molecular Formula C8H11BO2
Molecular Weight (g/mol) 149.98
MDL Number MFCD01631226
InChI Key VPRUMANMDWQMNF-UHFFFAOYSA-N
Synonym phenethylboronic acid, phenylethane boronic acid, peba, phenylethaneboronic acid, 2-phenyl-1-ethylboronic acid, 2-phenylethyl-1-boronic acid, boronic acid, 2-phenylethyl, 2-phenylethyl boronic acid, phenylethylboronic acid
PubChem CID 65389
SMILES OB(O)CCC1=CC=CC=C1

Specifications

Melting Point 82°C to 84°C
Quantity 5 g
IUPAC Name (2-phenylethyl)boronic acid
Formula Weight 149.98
Percent Purity 98%
Chemical Name or Material 2-Phenylethylboronic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.