missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylethanol, Extra Pure, SLR, Fisher Chemical

Product Code. 10375950 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Amber glass bottle
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10375950 500 mL Amber glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10375950 Supplier Fisher Chemical Supplier No. P/2526/08

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 60-12-8
Melting Point -27°C
Color Colorless
pH 6 to 7
Boiling Point 219°C
Packaging Amber glass bottle
Molecular Formula C8H10O
MDL Number MFCD00002886
Quantity 500 mL
Synonym phenethyl alcohol, benzeneethanol, phenylethyl alcohol, benzyl carbinol, phenylethanol, phenethanol, 2-phenylethyl alcohol, 2-phenyl-ethanol, beta-phenylethanol, 2-phenylethan-1-ol
InChI Key WRMNZCZEMHIOCP-UHFFFAOYSA-N
SMILES OCCC1=CC=CC=C1
IUPAC Name 2-phenylethanol
Molecular Weight (g/mol) 122.17
PubChem CID 6054
ChEBI CHEBI:49000
Formula Weight 122.17g/mol
Physical Form Liquid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Phenylethanol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.