missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylethanol, 98+%, Thermo Scientific Chemicals

Catalog Number 11442288
Click to view available options
Quantity:
250 g
1000 g
5000 g
This item is not returnable. View return policy
60-12-8
C8H10O
122.17
MFCD00002886
WRMNZCZEMHIOCP-UHFFFAOYSA-N
phenethyl alcohol, benzeneethanol, phenylethyl alcohol, benzyl carbinol, phenylethanol, phenethanol, 2-phenylethyl alcohol, 2-phenyl-ethanol, beta-phenylethanol, 2-phenylethan-1-ol
6054
CHEBI:49000
OCCC1=CC=CC=C1
This item is not returnable. View return policy
60-12-8
C8H10O
122.17
MFCD00002886
WRMNZCZEMHIOCP-UHFFFAOYSA-N
phenethyl alcohol, benzeneethanol, phenylethyl alcohol, benzyl carbinol, phenylethanol, phenethanol, 2-phenylethyl alcohol, 2-phenyl-ethanol, beta-phenylethanol, 2-phenylethan-1-ol
6054
CHEBI:49000
OCCC1=CC=CC=C1

2-Phenylethanol is used also as an aromatic essence and preservative in pharmaceutics and perfumery. It is also used as odor agents, air care products, cleaning and furnishing care products, laundry and dishwashing products, personal care products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Phenylethanol is used also as an aromatic essence and preservative in pharmaceutics and perfumery. It is also used as odor agents, air care products, cleaning and furnishing care products, laundry and dishwashing products, personal care products.

Solubility
Miscible with chloroform.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Melting Point °C to 27°C
Density 1.021
Boiling Point 219°C
Flash Point 101°C (214°F)
Assay Percent Range ≥98%
Linear Formula C6H5(CH2)2OH
Refractive Index 1.533
Quantity 250 g
UN Number UN2810
Beilstein 1905732
Merck Index 14,7224
Solubility Information Miscible with chloroform.
IUPAC Name 2-phenylethanol
Formula Weight 122.17
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material 2-Phenylethanol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'