missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylbutyric Acid, TRC

Product Code. 30006639
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006639 1 g 1g
17933435 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30006639 Supplier Toronto Research Chemicals Supplier No. TRCP3196001G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-27-7
Molecular Weight (g/mol) 164.2
IUPAC Name 2-phenylbutanoic acid
SMILES CCC(C(=O)O)c1ccccc1

Specifications

Molecular Formula C10 H12 O2
Quantity 1 g
Synonym (2RS)-2-Phenylbutanoic Acid, Benzeneacetic acid, α-ethyl-, Butyric acid, 2-phenyl- (8CI), α-Ethylbenzeneacetic acid, (RS)-2-Phenylbutanoic acid, (±)-2-Phenylbutanoic acid, (±)-2-Phenylbutyric acid, (±)-α-Ethylphenylacetic acid, (±)-α-Phenylbutyric acid, 2-Phenylbutanoic acid, 2-Phenylbutyric acid, NSC 1860, α-Ethyl-α-toluic acid, α-Ethylphenylacetic acid, α-Phenylbutyric acid
InChI Formula InChI=1S/C10H12O2/c1-2-9(10(11)12)8-6-4-3-5-7-8/h3-7,9H,2H2,1H3,(H,11,12)
Formula Weight 164.0837
Percent Purity >95%
Chemical Name or Material (2RS)-2-Phenylbutanoic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.