missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-2-Phenylbutyric acid, 98%, Thermo Scientific Chemicals

Product Code. 11470827
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11470827 100 g 100g
11480827 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11470827 Supplier Thermo Scientific Alfa Aesar Supplier No. B23222.22
  • ONLINE EXCLUSIVE PRICE
    £41.46
    £35.65 / 100g
    Save £5.81
    14% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 90-27-7 | Molecular Formula: C10H12O2 | Molecular Weight: 164.204 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-27-7
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00002667
InChI Key OFJWFSNDPCAWDK-UHFFFAOYSA-N
Synonym 2-phenylbutyric acid, alpha-ethylphenylacetic acid, butyric acid, 2-phenyl, alpha-phenylbutyric acid, benzeneacetic acid, .alpha.-ethyl, alpha-phenyl butyric acid, 2-phenylbutyricacid, benzeneacetic acid, alpha-ethyl, 2-phenylburyric acid, alpha-toluic acid, alpha-ethyl
PubChem CID 7012
ChEBI CHEBI:86545
IUPAC Name 2-phenylbutanoic acid
SMILES CCC(C1=CC=CC=C1)C(=O)O

Specifications

Melting Point 40°C to 44°C
Boiling Point 270°C to 272°C
Flash Point >110°C (230°F)
Refractive Index 1.515
Quantity 100 g
Beilstein 509876
Formula Weight 164.2
Percent Purity 98%
Chemical Name or Material (+/-)-2-Phenylbutyric acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.