missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylbenzothiazole, TRC

Product Code. 30138099
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138099 1 g 1g
30137462 500 mg 500mg
30005019 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138099 Supplier Toronto Research Chemicals Supplier No. TRCP3366351G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 883-93-2
Molecular Formula C13H9NS
Molecular Weight (g/mol) 211.28
Synonym 2-Phenylbenzothiazole, Benzothiazole, 2-phenyl-, 2-Phenylbenzothiazole, 2-Phenyl-1,3-benzothiazole, 2-Phenylbenzo[d]thiazole, NSC 1854, NSC 2034
IUPAC Name 2-phenyl-1,3-benzothiazole
SMILES c1ccc(cc1)c2nc3ccccc3s2

Tekniske data

Quantity 1 g
InChI Formula InChI=1S/C13H9NS/c1-2-6-10(7-3-1)13-14-11-8-4-5-9-12(11)15-13/h1-9H
Formula Weight 211.0456
Chemical Name or Material 2-Phenylbenzothiazole
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.