missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylbenzimidazole-5-sulfonic acid monohydrate, 98%

Product Code. 11436986
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11436986 5 g 5g
11446986 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11436986 Supplier Thermo Scientific Alfa Aesar Supplier No. B25307.06
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 27503-81-7 | C13H10N2O3S | 274.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27503-81-7
Molecular Formula C13H10N2O3S
Molecular Weight (g/mol) 274.29
MDL Number MFCD00053007
InChI Key UVCJGUGAGLDPAA-UHFFFAOYSA-N
Synonym ensulizole, 2-phenylbenzimidazole-5-sulfonic acid, phenylbenzimidazole sulfonic acid, 2-phenyl-1h-benzo d imidazole-5-sulfonic acid, 2-phenyl-1h-benzo d imidazole-6-sulfonic acid, 1h-benzimidazole-5-sulfonic acid, 2-phenyl, unii-9yq9di1w42, 2-phenyl-1h-benzimidazole-5-sulfonic acid, parsol hs, 2-phenyl-5-benzimidazolesulfonic acid
PubChem CID 33919
SMILES OS(=O)(=O)C1=CC=C2N=C(NC2=C1)C1=CC=CC=C1

Specifications

Melting Point >300°C
Quantity 5 g
Beilstein 922664
IUPAC Name 2-phenyl-3H-benzimidazole-5-sulfonic acid
Formula Weight 292.32 (274.30 Anhydrous)
Percent Purity 98%
Chemical Name or Material 2-Phenylbenzimidazole-5-sulfonic acid monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.