missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylazepane, TRC

Product Code. 30425661
Change view
Click to view available options
Quantity:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30425661 100 mg
30417809 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30425661 Supplier Toronto Research Chemicals Supplier No. TRCP319450100MG
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3466-82-8
Molecular Formula C12H17N
Molecular Weight (g/mol) 175.27
Synonym Hexahydro-2-phenyl-1H-azepine, 2-Phenyl-hexamethylenimine, 2-Phenylhexahydroazepine, 2-Phenylperhydroazepine
IUPAC Name 2-phenylazepane
SMILES C1CCNC(CC1)c2ccccc2

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C12H17N/c1-3-7-11(8-4-1)12-9-5-2-6-10-13-12/h1,3-4,7-8,12-13H,2,5-6,9-10H2
Formula Weight 175.1361
Chemical Name or Material 2-Phenylazepane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.