missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylacetohydrazide, TRC

Product Code. 30428564
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30428564 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30428564 Supplier Toronto Research Chemicals Supplier No. TRCP3210681G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Phenylacetohydrazide
CAS 937-39-3
Molecular Formula C8H10N2O
Quantity 1 g
InChI Formula InChI=1S/C8H10N2O/c9-10-8(11)6-7-4-2-1-3-5-7/h1-5H,6,9H2,(H,10,11)
SMILES NNC(=O)Cc1ccccc1
IUPAC Name 2-phenylacetohydrazide
Molecular Weight (g/mol) 150.18
Formula Weight 150.0793
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.