missing translation for 'onlineSavingsMsg'
Learn More

2-Phenyl-L-glycinamide hydrochloride, 97%, Thermo Scientific™

Product Code. p-7056681
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15462619

Brand: Thermo Scientific Alfa Aesar H65751.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60079-51-8
Molecular Formula C8H11ClN2O
Molecular Weight (g/mol) 186.639
MDL Number MFCD00270568
InChI Key LXOAFHGMXCFTOU-FJXQXJEOSA-N
Synonym s-2-amino-2-phenylacetamide hydrochloride, l-phenylglycine amide hydrochloride, h-phg-nh2 hcl, h-phg-nh2.hcl, 2s-2-amino-2-phenylacetamide hydrochloride, hpphg-nh2 hcl, h-phg-nh2 . hcl, phenylglycine amide hydrochloride, s-2-amino-2-phenyl-acetamide hydrochloride, alphas-alpha-amino-benzeneacetamide hydrochloride
PubChem CID 22850726
IUPAC Name (2S)-2-amino-2-phenylacetamide;hydrochloride
SMILES C1=CC=C(C=C1)C(C(=O)N)N.Cl

Specifications

Quantity 1 g
Beilstein 4823667
Formula Weight 186.64
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Phenyl-L-glycinamide hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.