missing translation for 'onlineSavingsMsg'
Learn More

2-Phenyl-3-butyn-2-ol, 98%

Product Code. 12659651 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass bottle
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
12659651 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12659651 Supplier Thermo Scientific Acros Supplier No. 199691000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 127-66-2 | C10H10O | 146.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 127-66-2
Melting Point 50.0°C to 52.0°C
Color White
Boiling Point 102.0°C to 103.0°C (12.0 mmHg)
Flash Point 96°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Molecular Formula C10H10O
Linear Formula HC≡CC(C6H5)(OH)CH3
Quantity 100 g
Beilstein 06,II,559
Merck Index 10,7158
Synonym 2-phenyl-3-butyn-2-ol, methylphenylethynylcarbinol, 3-hydroxy-3-phenyl-1-butyne, 3-butyn-2-ol, 2-phenyl, 3-phenyl-butin-1-ol-3, alpha-ethynyl-alpha-methylbenzyl alcohol, alpha-ethynyl-alpha-methylbenzenemethanol, benzenemethanol, .alpha.-ethynyl-.alpha.-methyl, 3-phenyl-butin-1-ol-3 german, benzenemethanol, alpha-ethynyl-alpha-methyl
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in acetone,benzene and most other common,organic solvents
InChI Key KSLSOBUAIFEGLT-UHFFFAOYSA-N
SMILES CC(C#C)(C1=CC=CC=C1)O
IUPAC Name 2-phenylbut-3-yn-2-ol
Molecular Weight (g/mol) 146.19
PubChem CID 92098
Formula Weight 146.19
Percent Purity 98%
Physical Form Crystals
Chemical Name or Material 2-Phenyl-3-butyn-2-ol, 98+%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Phenyl-3-butyn-2-ol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and any thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.