missing translation for 'onlineSavingsMsg'
Learn More

2-Phenyl-2-propanol, 99%

Product Code. 10133280 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10133280 25 g Glass bottle 25g
10735611 5 g Glass bottle 5g
10475485 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10133280 Supplier Thermo Scientific Acros Supplier No. 130830250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 617-94-7 | C9H12O | 136.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 617-94-7
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.19
MDL Number MFCD00004456
InChI Key BDCFWIDZNLCTMF-UHFFFAOYSA-N
Synonym 2-phenyl-2-propanol, dimethylphenylcarbinol, 1-hydroxycumene, dimethylphenylmethanol, alpha-cumyl alcohol, phenyldimethylcarbinol, 2-propanol, 2-phenyl, 2-phenylisopropanol, alpha,alpha-dimethylbenzyl alcohol, 2-phenyl-propan-2-ol
PubChem CID 12053
IUPAC Name 2-phenylpropan-2-ol
SMILES CC(C)(O)C1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 32°C to 34°C
Color Colorless to Yellow
Density 0.9730g/mL
Boiling Point 88°C to 90°C (11.0 mmHg)
Flash Point 88°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5C(CH3)2OH
Refractive Index 1.5190 to 1.522
Quantity 25 g
Beilstein 06, 506
Specific Gravity 0.973
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol,ether,benzene
Formula Weight 136.19
Percent Purity 99%
Physical Form Liquid After Melting
Chemical Name or Material 2-Phenyl-2-propanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.