missing translation for 'onlineSavingsMsg'
Learn More

2-Phenyl-1-propylboronic acid pinacol ester, 97%, Thermo Scientific™

Catalog Number 15467635
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
280559-30-0
C15H23BO2
246.157
MFCD09953475
GTZNZJQZUWIDTN-UHFFFAOYSA-N
2-phenyl-1-propylboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-2-phenylpropyl-1,3,2-dioxaborolane, 2-phenylpropylboronic acid pinacol ester, 2-2-phenylpropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
46739058
4,4,5,5-tetramethyl-2-(2-phenylpropyl)-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)CC(C)C2=CC=CC=C2
This item is not returnable. View return policy
280559-30-0
C15H23BO2
246.157
MFCD09953475
GTZNZJQZUWIDTN-UHFFFAOYSA-N
2-phenyl-1-propylboronic acid pinacol ester, 4,4,5,5-tetramethyl-2-2-phenylpropyl-1,3,2-dioxaborolane, 2-phenylpropylboronic acid pinacol ester, 2-2-phenylpropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
46739058
4,4,5,5-tetramethyl-2-(2-phenylpropyl)-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)CC(C)C2=CC=CC=C2

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 246.16
Percent Purity 97%
Chemical Name or Material 2-Phenyl-1-propylboronic acid pinacol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'