missing translation for 'onlineSavingsMsg'
Learn More

2-Phenyl-1,3-thiazole-4-carbaldehyde, ≥90%, Maybridge™

Brand:  Maybridge CC18304CB

Code : 31

Additional Details : CAS Number : 20949-81-9 Weight : 0.05025kg

Packaging Amber Glass Bottle
Quantity 250mg

 View more versions of this product

Product Code. 10049211

  • £21.00 / 250mg

Estimated Shipment: 13-10-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

20949-81-9
189.232
OLLKCCGWRITPOV-UHFFFAOYSA-N
736524
C1=CC=C(C=C1)C2=NC(=CS2)C=O
C10H7NOS
MFCD02681934
2-phenylthiazole-4-carbaldehyde, 4-thiazolecarboxaldehyde, 2-phenyl, 2-phenyl-4-formylthiazole, 4-formyl-2-phenyl-1,3-thiazole, 4-thiazolecarboxaldehyde,2-phenyl, 2-phenyl-thiazole-4-carbaldehyde, 2-phenyl-4-thiazolecarboxaldehyde, 4-formyl-1,3-thiazol-2-yl benzene, 2-phenyl-1,3-thiazole-4-carboxaldehyde
2-phenyl-1,3-thiazole-4-carbaldehyde

Specifications

2-Phenyl-1, 3-thiazole-4-carbaldehyde
250mg
94.5
95%
MFCD02681934
OLLKCCGWRITPOV-UHFFFAOYSA-N
2-phenyl-1,3-thiazole-4-carbaldehyde
736524
95%
Amber Glass Bottle
20949-81-9
100.0
C10H7NOS
2-phenylthiazole-4-carbaldehyde, 4-thiazolecarboxaldehyde, 2-phenyl, 2-phenyl-4-formylthiazole, 4-formyl-2-phenyl-1,3-thiazole, 4-thiazolecarboxaldehyde,2-phenyl, 2-phenyl-thiazole-4-carbaldehyde, 2-phenyl-4-thiazolecarboxaldehyde, 4-formyl-1,3-thiazol-2-yl benzene, 2-phenyl-1,3-thiazole-4-carboxaldehyde
C1=CC=C(C=C1)C2=NC(=CS2)C=O
189.232
189.24
Videos
SDS Documents
Certificates