missing translation for 'onlineSavingsMsg'
Learn More

(2-Phenyl-1,3-thiazol-4-yl)methylamine, 97%, Thermo Scientific™

Product Code. 10356304
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10356304 10 g Amber Glass Bottle 10g
10079231 1 g Amber Glass Bottle 1g
10041124 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10356304 Supplier Thermo Scientific Maybridge Supplier No. CC18313EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 165736-03-8
Molecular Formula C10H10N2S
Molecular Weight (g/mol) 190.264
MDL Number MFCD02682055
InChI Key PMFCMLGAPHZRTE-UHFFFAOYSA-N
Synonym 2-phenyl-1,3-thiazol-4-yl methylamine, 2-phenylthiazol-4-yl methanamine, 2-phenyl-1,3-thiazol-4-yl methanamine, c-2-phenyl-thiazol-4-yl-methylamine, 1-2-phenyl-1,3-thiazol-4-yl methanamine, 4-thiazolemethanamine, 2-phenyl, 4-aminomethyl-2-phenyl-1,3-thiazole, enamine_005991, 2-phenyl-1,3-thiazol-4-yl methanamine dihydrochloride, chembrdg-bb 4101065
PubChem CID 736526
IUPAC Name (2-phenyl-1,3-thiazol-4-yl)methanamine
SMILES C1=CC=C(C=C1)C2=NC(=CS2)CN

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 190.27
Percent Purity 97%
Chemical Name or Material (2-Phenyl-1, 3-thiazol-4-yl)methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.