missing translation for 'onlineSavingsMsg'
Learn More

2-Phenoxynicotinic acid, 98%

Product Code. 11423594
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11423594 5 g 5g
11389559 1 g 1g
11433594 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11423594 Supplier Thermo Scientific Alfa Aesar Supplier No. A11247.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 35620-71-4 | C12H9NO3 | 215.208 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35620-71-4
Molecular Formula C12H9NO3
Molecular Weight (g/mol) 215.208
MDL Number MFCD00014629
InChI Key CQGAXJGXGLVFGJ-UHFFFAOYSA-N
Synonym 2-phenoxynicotinic acid, 3-pyridinecarboxylic acid, 2-phenoxy, 2-phenoxy-3-pyridinecarboxylic acid, 2-phenoxy-nicotinic acid, acide 2-phenoxy nicotinique french, acide 2-phenoxy nicotinique, acmc-20a2ql, maybridge1_000744, cambridge id 5832716, 2-phenoxy nicotinic acid
PubChem CID 123450
IUPAC Name 2-phenoxypyridine-3-carboxylic acid
SMILES C1=CC=C(C=C1)OC2=C(C=CC=N2)C(=O)O

Specifications

Melting Point 179°C to 182°C
Quantity 5 g
Beilstein 172906
Formula Weight 215.21
Percent Purity 98%
Chemical Name or Material 2-Phenoxynicotinic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.