missing translation for 'onlineSavingsMsg'
Learn More

2-Phenoxyethylamine, ≥95%, Thermo Scientific™

Product Code. 10342065
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10342065 5 g 5g
10171032 1 g 1g
10352065 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10342065 Supplier Thermo Scientific Maybridge Supplier No. RF03254DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 1758-46-9
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.182
InChI Key IMLAIXAZMVDRGA-UHFFFAOYSA-N
Synonym 2-phenoxyethylamine, phenoxyethylamine, ethanamine, 2-phenoxy, 2-phenoxylethylamine, 2-phenoxy-ethylamine, 2-aminoethoxy benzene, ethylamine, 2-phenoxy, alpha-phenoxy-beta-aminoethane, unii-8dgq1b38r5, 2-phenoxyethylamino
PubChem CID 15651
IUPAC Name 2-phenoxyethanamine
SMILES C1=CC=C(C=C1)OCCN

Specifications

CAS Min % 95.0
Density 1.0480g/mL
Assay Percent Range 95%
Linear Formula C6H5O(CH2)2NH2
Quantity 5 g
Formula Weight 137.18
Percent Purity 95%
Chemical Name or Material 2-Phenoxyethylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.