missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Phenoxyethyl-1,1,2,2-d4 Alcohol, TRC
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Chemical Name or Material | 2-(Phenoxy)ethanol-1,1,2,2-D4 |
| CAS | 1219804-65-5 |
| Molecular Formula | C8H6D4O2 |
| Quantity | 50 mg |
| Synonym | 2-Phenoxyethyl-1,1,2,2-d4 Alcohol |
| InChI Formula | InChI=1S/C8H10O2/c9-6-7-10-8-4-2-1-3-5-8/h1-5,9H,6-7H2/i6D2,7D2 |
| SMILES | OC([2H])(C([2H])(OC1=CC=CC=C1)[2H])[2H] |
| IUPAC Name | 2-phenoxyethan-1,1,2,2-d4-1-ol |
| Molecular Weight (g/mol) | 142.19 |
| Formula Weight | 142.09 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction