missing translation for 'onlineSavingsMsg'
Learn More

2-Phenoxyethanol, 94%

Product Code. 11429702
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11429702 1000 g 1000g
11439702 250 g 250g
11449702 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11429702 Supplier Thermo Scientific Alfa Aesar Supplier No. A10786.0B
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 122-99-6 | C8H10O2 | 138.166 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
2-Phenoxyethanol is used as a solvent for cellulose acetate, inks and resins. It is also used as a fixative for perfumes, insect repellents and topical antiseptics. It is utilized as a surface active agent. It acts as an anesthetic in the aquaculture of some fish. Further, it is used as a preservative and cosmetic raw material. In addition to this, it is used in preparation of active pharmaceutical ingredients and dyes.

Solubility
Miscible with water, ethanol, propanol and propylene glycol.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-99-6
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.166
MDL Number MFCD00002857
InChI Key QCDWFXQBSFUVSP-UHFFFAOYSA-N
Synonym phenoxyethanol, ethylene glycol monophenyl ether, phenyl cellosolve, ethanol, 2-phenoxy, phenoxytol, phenoxethol, phenoxetol, ethylene glycol phenyl ether, phenoxyethyl alcohol, 1-hydroxy-2-phenoxyethane
PubChem CID 31236
ChEBI CHEBI:64275
IUPAC Name 2-phenoxyethanol
SMILES C1=CC=C(C=C1)OCCO

Specifications

Melting Point 11°C to 14°C
Density 1.102
Boiling Point 245°C
Flash Point 121°C (249°F)
Assay Percent Range 94%
Linear Formula C6H5O(CH2)2OH
Refractive Index 1.537
Quantity 1000 g
Beilstein 1364011
Merck Index 14,7257
Solubility Information Miscible with water,ethanol,propanol and propylene glycol.
Formula Weight 138.17
Percent Purity 94%
Physical Form Liquid
Chemical Name or Material 2-Phenoxyethanol
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.