missing translation for 'onlineSavingsMsg'
Learn More

2-Pentylidenecyclohexanone, TRC

Product Code. 17953645
Change view
Click to view available options
Quantity:
500 mg
5 g
Unit Size:
500mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17953645 500 mg 500mg
17934735 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17953645 Supplier Toronto Research Chemicals Supplier No. TRCP280255500MG
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25677-40-1
Molecular Formula C11 H18 O
Molecular Weight (g/mol) 166.26
Synonym 2-Pentylidenecyclohexanone
IUPAC Name (2E)-2-pentylidenecyclohexan-1-one
SMILES CCCC\C=C\1/CCCCC1=O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C11H18O/c1-2-3-4-7-10-8-5-6-9-11(10)12/h7H,2-6,8-9H2,1H3/b10-7+
Formula Weight 166.1358
Chemical Name or Material 2-Pentylidenecyclohexanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.