missing translation for 'onlineSavingsMsg'
Learn More

2-Pentylfuran, TRC

Product Code. 30136553
Change view
Click to view available options
Quantity:
500 mg
1 g
10 g
Unit Size:
10g
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136553 10 g 10g
17971225 1 g 1g
17923035 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136553 Supplier Toronto Research Chemicals Supplier No. TRCP28495310G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3777-69-3
Molecular Formula C9 H14 O
Molecular Weight (g/mol) 138.21
Synonym Furan, 2-pentyl-, 2-Pentylfuran, 2-Amylfuran, 2-n-Pentylfuran, Dihydro-5-pentyl-2(hydro)-furan
IUPAC Name 2-pentylfuran
SMILES CCCCCc1occc1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C9H14O/c1-2-3-4-6-9-7-5-8-10-9/h5,7-8H,2-4,6H2,1H3
Formula Weight 138.1045
Percent Purity >95%
Chemical Name or Material 2-Pentylfuran
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.